


Hexane (mixture of isomers) ≥95%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
BJ650552-1L
:
- Pack type:Glass bottle
- Size:1 L
- Cat. No.:BJ650552-1L
- Supplier no.:650552-1L
- Vapor Pressure:256 mmHg ( 37.7 °C)
- Assay (GC):Min. 95.0 %
- Non-volatile matter:Max. 0.0001 %
- Water (Karl Fischer):Max. 0.01 %
- Refractive index (n 20/D):1.373 - 1.377
- Absorbance at 400 nm:Max. 0.005
- Absorbance 280 nm:Max. 0.005
- Absorbance at 250 nm:Max. 0.01
- Absorbance at 220 nm:Max. 0.05
- Absorbance at 210 nm:Max. 0.20
- Absorbance at 200 nm:Max. 0.50
- Absorbance at 195 nm:Max. 1.0
- Fluorescence (chinin) at 365 nm:Max. 1 ppb
- IR spectrum:complying
- GC (as heptachlor epoxide):Max. 10 ng/l
- Autoignition Temperature:453 °F
- Refractive Index:n20/D 1.375 (lit.)
:
:86.18 g/mol
:66…69 °C(1.013 hPa)
:–95 °C
:0.660…0.668 g/cm³ (20 °C)
:7.4 %(V)
:Ambient
:203-777-6
:1208
:3,II
:GHS07
:GHS08
:GHS02
:110-54-3
:92112-69-1
:H₃C(CH₂)₄CH₃
:MFCD02179311