


n-Heptane 99%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Catalog # BJ650536-1L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:1 L
- Cat. No.:BJ650536-1L
- Supplier no.:650536-1L
Specifications
Specification Test Results
- Autoignition Temperature:433 °F
- Purity (GC):Min. 98.5 %
- IR spectrum:complying
- Refractive index (n 20/D):1.3850 - 1.3890
- Non-volatile matter:Max. 0.0001 %
- Water (Karl Fischer):Max. 0.01 %
- Absorbance at 400 nm:Max. 0.005
- Absorbance at 300 nm:Max. 0.005
- Absorbance at 250 nm:Max. 0.01
- Absorbance at 225 nm:Max. 0.10
- Absorbance at 200 nm:Max. 1.0
- Fluorescence (chinin) at 365 nm:Max. 1.0 ppb
- GC (as heptachlor epoxide):Max. 2 µg/l
- Vapor Density:3.5 (vs air)
- Vapor Pressure:83 mmHg ( 37.7 °C)
- Explosion Limit:7 %
Specification Test Results
About this item
:
:100.20 g/mol
Boiling Pt:98.2…98.4 °C (1.000 hPa)
Melting Pt:–90.5 °C
Density:0.69 g/cm³ (15 °C)
Flash Pt:6.7 %(V)
Storage Temperature:Ambient
EINECS:205-563-8
UN:1206
ADR:3,II
:GHS07
:GHS08
:GHS02
CAS Number:142-82-5
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544