

Tetrachloroethylene ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Catalog # BJ270393-1L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:1 L
- Cat. No.:BJ270393-1L
- Supplier no.:270393-1L
Specifications
Specification Test Results
- Identity (IR):complying
- Assay (GC):Min. 99.90 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.05 %
- Absorbance at 400 nm:Max. 0.03
- Absorbance at 350 nm:Max. 0.05
- Absorbance at 305 nm:Max. 0.10
- Absorbance at 300 nm:Max. 0.20
- Absorbance at 295 nm:Max. 0.30
- Absorbance at 290 nm:Max. 1.0
- APHA:Max. 10
- Vapor Pressure:13 mmHg ( 20 °C)
- Vapor Density:5.83 (vs air)
- Refractive Index:n20/D 1.505 (lit.)
Specification Test Results
About this item
:
:165.83 g/mol
Boiling Pt:120-122 °C (1.013 hPa)
Boiling Pt:120…122 °C (1.013 hPa)
Melting Pt:–22 °C
Density:1.622…1.624 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-825-9
UN:1897
ADR:6.1,III
:GHS07
:GHS08
CAS Number:127-18-4
Formula:C₂Cl₄
MDL Number:MFCD00000834