


N,N-Dimethylformamide ≥99.9%, CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:68-12-2
:
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284
:73.09 g/mol
Boiling Pt:152…153 °C (1.013 hPa)
Melting Pt:–61 °C
Density:0.94 g/cm³ (20 °C)
Flash Pt:57.5 °C (1.013.25 hPa)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
:GHS02
:GHS08
:GHS07
Specification Test Results
- Suitability for the GC Headspace:complying
- Assay (GC):Min. 99.9 %
- Refractive index (n 20/D):1.429 - 1.431
- Water (Karl Fischer):Max. 0.03 %
- Absorbance at 270 nm:Max. 0.700
- Absorbance at 275 nm:Max. 0.250
- Absorbance at 300 nm:Max. 0.070
- Absorbance at 320 nm:Max. 0.022
Specification Test Results
Related Information
Frequently Bought Together