Order Entry
United States
ContactUsLinkComponent
Dimethyl sulfoxide ≥99.9%, CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Dimethyl sulfoxide ≥99.9%, CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Catalog # BJ51779-1L
CAS Number:  
undefined
Dimethyl sulfoxide ≥99.9%, CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Catalog # BJ51779-1L
Supplier Number:  51779-6X1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    BJ51779-1L
  • Supplier no.:
    51779-1L

Specifications

Specification Test Results

  • Suitability for the GC Headspace:
    complying
  • Assay (GC):
    min. 99.9 %
  • Refractive index (n 20/D):
    1.477 - 1.480
  • Water (Karl Fischer):
    max. 0.04 %
  • Absorbance at 268 nm:
    max. 0.522
  • Absorbance at 275 nm:
    max. 0.221
  • Absorbance at 300 nm:
    max. 0.070
  • Absorbance at 350 nm:
    max. 0.022
  • Absorbance at 400 nm:
    max. 0.008

Specification Test Results

About this item

 :
 :78.14 g/mol
Boiling Pt:189 °C
Melting Pt:18.5 °C
Density:1.104 g/cm³ (20 °C)
Flash Pt:28.5 %(V)
Storage Temperature:Ambient
EINECS:200-664-3
CAS Number:67-68-5
Formula:(CH₃)₂SO
MDL Number:MFCD00002089