1-Propanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:71-23-8
:
Formula:H₃CCH₂CH₂OH
MDL Number:MFCD00002941
:60.1 g/mol
Boiling Pt:96…98 °C (1.013 hPa)
Melting Pt:–127 °C
Density:0.803…0.804 g/cm³ (20 °C)
Flash Pt:10.8 %(V)
Storage Temperature:Ambient
EINECS:200-746-9
UN:1274
ADR:3,II
:GHS07
:GHS05
:GHS02
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (GC):Min. 99.9 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.05 %
- Free acid (as C₂H₅COOH):Max. 0.001 %
- Transmittance at 220 nm:Min. 40 %
- Transmittance at 240 nm:Min. 85 %
- Transmittance from 275 nm:Min. 99 %
- Vapor Pressure:14.9 mmHg ( 20 °C)
- Autoignition Temperature:700 °F
- Vapor Density:2.1 (vs air)
- Refractive Index:n20/D 1.384 (lit.)
Specification Test Results
Related Information
Frequently Bought Together