


n-Pentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:109-66-0
:
Formula:H₃C(CH₂)₃CH₃
MDL Number:MFCD00009498
:72.15 g/mol
Boiling Pt:36…37 °C
Melting Pt:–131 °C
Density:0.631 g/cm³ (15 °C) (lit.)
Flash Pt:<–35 °C
Storage Temperature:Ambient
EINECS:203-692-4
UN:1265
ADR:3,II
:GHS07
:GHS08
:GHS02
Specification Test Results
- Assay (GC):Min. 99 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.01 %
- Suitability for residue analysis:Max. 5 ng/l
- Explosion Limit:~8.3 %
- Autoignition Temperature:500 °F
Specification Test Results
Related Information
Frequently Bought Together