


n-Hexane ≥95% (by GC), CHROMASOLV™ for pesticide residue analysis, for analysis of dioxins, furans and PCB, Riedel-de Haën™
Catalog # BJ34412-2.5L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:2.5 L
- Cat. No.:BJ34412-2.5L
- Supplier no.:34412-2.5L
Specifications
Specification Test Results
- Assay (GC):Min. 95 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.01 %
- Suitability for GC-MS of dioxines, furanes, PCPs:complying
- Vapor Density:~3 (vs air)
- Vapor Pressure:~132 mmHg ( 20 °C)
- Refractive Index:n20/D 1.375 (lit.)
- Explosion Limit:7.7 %
Specification Test Results
About this item
:
:86.18 g/mol
Boiling Pt:66…69 °C(1.013 hPa)
Melting Pt:–95 °C
Density:0.660…0.668 g/cm³ (20 °C)
Flash Pt:7.4 %(V)
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
:GHS07
:GHS08
:GHS02
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520