Dichloromethane ≥99.8% (by GC) stabilized, CHROMASOLV™ for pesticide residue analysis, for analysis of dioxins, furans and PCB, Riedel-de Haën™
Catalog # BJ34411-2.5L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:2.5 L
- Cat. No.:BJ34411-2.5L
- Supplier no.:34411-2.5L
Specifications
Specification Test Results
- Refractive Index:n20/D 1.424 (lit.)
- Assay (GC):Min. 99.8 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.02 %
- Suitability for GC-MS of dioxines, furanes, PCPs:complying
- Vapor Pressure:24.45 psi ( 55 °C)
- Autoignition Temperature:1223 °F
- Contains:~25 mg/L amylene as stabilizer
Specification Test Results
About this item
:
:84.93 g/mol
Boiling Pt:39…41 °C (1.013 hPa)
Melting Pt:–97 °C
Density:1.32…1.33 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-838-9
UN:1593
ADR:6.1,III
:GHS08
:GHS07
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881