

Formic acid 49 - 51% (by titrimetric analysis) in water, Puriss. p.a. for HPLC, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:64-18-6
:
Formula:HCO₂H
MDL Number:MFCD00003297
:46.03
Boiling Pt:101 °C (1.013 hPa.) (anhydr.)
Melting Pt:4 °C (anhydr.)
Density:1.22 g/cm3
Flash Pt:49.5 °C
Storage Temperature:Ambient
UN:3412
ADR:8,II
:GHS07
:GHS05
Specification Test Results
- Assay:49.0 - 51.0 %
- Absorbance at 270 nm:Max. 0.05
- Absorbance 280 nm:Max. 0.04
- Absorbance at 290 nm:Max. 0.03
- Absorbance at 300 nm:Max. 0.02
- Absorbance 500 nm:Max. 0.02
Specification Test Results
Frequently Bought Together