Silver nitrate 1.0 M AgNO3 (1.0N) volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:7761-88-8
:
Formula:AgNO₃
MDL Number:MFCD00003414
:169.87 g/mol
Boiling Pt:100 °C
Density:1.140 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3264
ADR:8,III
:GHS05
Specification Test Results
- Appearance:complying
- Coloration:complying
- Clearness:complying
- Assay titr.:0.950 - 1.050 mol/l
- Traceable to NIST SRM:complying
- BAM/EMPA reference material:complying
- BAM/EMPA Certificate Number:complying
Specification Test Results
Related Information
Frequently Bought Together