ContactUsLinkComponent
Potassium hydroxide 0.1 M (0.1 N) in methanol ≥99% volumetric solution, Fluka™
Health
Flammable
Potassium hydroxide 0.1 M (0.1 N) in methanol ≥99% volumetric solution, Fluka™
Certificates
Health
Flammable
Potassium hydroxide 0.1 M (0.1 N) in methanol ≥99% volumetric solution, Fluka™
Specification Test Results
  • Assay:
    0.0950 - 0.1050 N
  • Carbonate (as K₂CO₃):
    Max. 0.003 N
  • Traceable to NIST SRM:
    complying