EDTA disodium salt 0.05 M EDTA-Na2 volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:139-33-3
:
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672
:336.21 g/mol
Density:1.01 g/cm³
Storage Temperature:Ambient
Specification Test Results
- Assay:0.0490 - 0.0510 mol/l
- Traceable to NIST SRM:effective
Specification Test Results
Frequently Bought Together