Order Entry
United States
ContactUsLinkComponent
Ammonia solution 5.0 M NH4OH (5.0N) volumetric solution, Fluka™
Irritant
Corrosive
Ammonia solution 5.0 M NH4OH (5.0N) volumetric solution, Fluka™
Certificates
Irritant
Corrosive
Ammonia solution 5.0 M NH4OH (5.0N) volumetric solution, Fluka™
Certificates

Specification Test Results

  • Assay:
    4.90 - 5.10 N
  • Traceable to NIST SRM:
    effective
  • Explosion Limit:
    27 %

Specification Test Results

Related Information

Frequently Bought Together