Whole-Cell Pipette Perfusion Kit, ALA Scientific Instruments
This kit is used for the internal pipette perfusion and capable of sensitive and precise vacuum and pressure control of the pipette and designed with a stable mechanism for solution exchange.
- Perfusion pipette holder
- Pressure and vacuum controller to balance pressure on patch while driving fluid flow
- Quartz capillary tube provides optimal noise reduction and durability for proximal positioning to pipette tip
Frequently Bought Together