COSFect™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
Certificates
About this item
COSFect is a lipid based transfection reagent specifically developed for COS cell transfection.
- Highly efficient with COS cell lines
- Ready-to-use: no need for additional buffer
- Low nucleic acid amount - minimized toxicity
- High level of nucleic acid compaction
- Easy and straightforward protocol
- Compatible with any culture medium: medium changed not required
The cationic design of COSFect reagent allows high nucleic acid compaction for a high transfection efficiency into COS cells.
Delivery: Shipped at room temperature
Packaging: Store at -20 °C
Specifications
- Cell Specific:COS
- DNA:Yes
- dsRNA:Yes
- miRNA:Yes
- mRNA:Yes
- shRNA:Yes
- siRNA:Yes
Specifications
Frequently Bought Together