PolyMag™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
Certificates
About this item
Based in the Magnetofection technology, this reagent is composed of magnetic nanoparticles coated with cationic molecules.
- Highly efficient for primary cells
- Powerful for hard to transfect cells
- Non toxic
- Serum-compatible
- Simple, rapid and easy-to-use
- For all types of nucleic acids
It is a versatile reagent ideal for transfecting primary cells and hard-to-transfect cells.
Accessories information: This reagent needs to be used with a magnetic plate
Delivery information: Shipped at room temperature
Packaging: Store at +4°C
Specifications
- Cell Specific:Mammalian, Adherent
- DNA:Yes
- dsRNA:Yes
- miRNA:Yes
- mRNA:Yes
- shRNA:Yes
- siRNA:Yes
Specifications
Frequently Bought Together