Order Entry
United States
ContactUsLinkComponent
Calcium Phosphate (CaPo) Transfection Kit, OZ Biosciences
Calcium Phosphate (CaPo) Transfection Kit, OZ Biosciences
Catalog # 103258-268
Supplier:  OZ Biosciences
CAS Number:  
undefined
Calcium Phosphate (CaPo) Transfection Kit, OZ Biosciences
Catalog # 103258-268
Supplier:  OZ Biosciences
Supplier Number:  CP90000
CAS Number:  

Specifications

  • Bioproduction:
    Yes
  • Cell Specific:
    HEK 293, Mammalian
  • Description:
    Calcium Phosphate (CaPo) Transfection Kit
  • Size:
    4 x 15 mL Hepes Buffered Saline + 3.5 mL CaCl₂
  • Cat. No.:
    103258-268
  • No. of reactions:
    100
  • Supplier no.:
    CP90000

Specifications

About this item

The CaPO transfection kit is simple and easy to use.

  • Ideal for virus production
  • High HEK 293 cells transfection efficiency
  • Suitable for producing recombinant proteins
  • Serum compatible
  • Simple, Ready-to-use and Rapid

It allows reaching between 95 and 100% of HEK 293 transfected cells and a very high titer for virus production.

Delivery: Shipped at room temperature
Packaging: Store at -20 °C