3D-Fect™ Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
Certificates
About this item
3D-Fect is specifically developed to directly transfect cells cultured in 3D scaffolds.
- Highly efficient: cell lines and primary cells
- Specific for 3D Scaffolds
- Compatible with all types of nucleic acids
- Completely biodegradable
- Long term protein expression
- Serum Compatible
This 3D Transfection Reagent is suitable for all kinds of scaffolds and cells.
Delivery: Shipped at room temperature
Packaging: Store at +4 °C
Specifications
- DNA:Yes
- dsRNA:Yes
- mRNA:Yes