Sodium laureth sulfate 27% in aqueous solution
Catalog # 700015-838
Supplier: Avantor
Small Business Enterprise
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Drum
- Size:200 L
- Cat. No.:700015-838
- Supplier no.:S2139-200LTB
Specifications
About this item
:
Density:1.000
Storage Temperature:Ambient
CAS Number:9004-82-4
Formula:(C₂H₄O)x·C₁₂H₂₆O₄S·Na