ContactUsLinkComponent
Denoyer-Geppert® Spinal Palpation Trainer
Denoyer-Geppert® Spinal Palpation Trainer
Catalog # 470225-910
undefined
Denoyer-Geppert® Spinal Palpation Trainer
Catalog # 470225-910
Supplier Number:  SP672
Specifications
  • Description:
    Spine Palpation
  • Cat. No.:
    470225-910
  • Supplier no.:
    SP672
Specifications
About this item
The palpation training kit includes two levels of tissue simulation to inform the varying levels of soft tissue that exists throughout the general population and spine.
  • Practice palpation through 2 thicknesses of tissue.
This new spine palpation trainer simulates the soft tissue (muscle, skin, and fat) that overlies the vertebral column, allowing the user to improve their tactile and kinesthetic skills at knowing which vertebral level and which spinous process is being palpated. Perfect for practitioners and students of massage, physical therapy, chiropractic's, osteopathic, or orthopedics.