

cis-Platin ≥98% (by HPLC), Calbiochem®
Catalog # 80501-024
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:50 mg
- Cat. No.:80501-024
- Supplier no.:232120-50MG
Specifications
Specification Test Results
- Appearance:Yellow solid
- Purity (by HPLC):≥98%
- Solubility:DMSO (10 mg/mL), H₂O with 154 mM NaCl (1 mg/mL), or PBS with 140 mM NaCl (1 mg/mL)
- Storage:Store at 2 to 8 °C
- Stability:Unstable in solution; reconstitute just prior to use.
Specification Test Results
About this item
:
:300.05 g/mol
Merck Index:13,02341
Density:3.738 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:239-733-8
UN:3288
ADR:6.1,II
:GHS06
:GHS08
CAS Number:15663-27-1
Formula:H₆Cl₂N₂Pt
MDL Number:MFCD00011623