DOG1 IHC Positive Control Slides, Newcomer Supply
Supplier: Newcomer Supply
Certificates
About this item
DOG1 control slides contain sections of positive staining gastrointestinal stromal tumor (GIST) and negative staining kidney.
- Tissue type: (+) GIST / (-) Kidney
- All control tissue is sectioned on Superfrost® Plus slides
- Staining validation technical memo included in each set
- All tissues are formalin fixed, paraffin embedded FFPE
- All slides are air dried
- IHC slides are cut at 4 µm
- Standard tissue placement is mid-high on slide
Newcomer supply DOG1 control slides are for the positive immunohistochemical staining of DOG1, expressed predominantly on the plasma membrane of gastrointestinal stromal tumors (GIST).
Staining Results:
DOG1 positive expression will stain brown membrane & cytoplasmic staining.
Kidney will stain negative.
Nuclei will stain blue.
These positive control slides are intended to be used to verify histological techniques and reagent reactivity. These slides are to be used for the qualitative purpose of determining positive or negative results, and are not intended to be used for any quantitative purpose.
Ordering information: All controls are cut in serial sections.
10 set = 10 unstained slides and 1 stained slide.
98 set = 98 unstained slides and 2 stained slides. (First & Last Slides are Stained)
Reactivity of IHC control slides are guaranteed for one year from the date of receipt and should be revalidated after one year to verify continued reactivity.
Please note the following: Allow 2-3 weeks for delivery for large sets. In human tissue unless otherwise indicated.
10 set = 10 unstained slides and 1 stained slide.
98 set = 98 unstained slides and 2 stained slides. (First & Last Slides are Stained)
Reactivity of IHC control slides are guaranteed for one year from the date of receipt and should be revalidated after one year to verify continued reactivity.
Please note the following: Allow 2-3 weeks for delivery for large sets. In human tissue unless otherwise indicated.
Frequently Bought Together