BCL2 Positive Control Slides, IHC Newcomer Supply
Catalog # 75876-836
Supplier: Newcomer Supply
Specifications
- Species:
- Description:ICH control slide, Bcl2
- Environmentally Preferable:
- Cat. No.:75876-836
- Packaging:10/Set
- Supplier no.:3052A
Specifications
About this item
BCL2 control slides contain a section of positive staining lymphoma.
- Tissue type: (+) Lymphoma
- All control tissue is sectioned on Superfrost® Plus slides
- Staining validation technical memo included in each set
- All tissues are formalin fixed, paraffin embedded FFPE
- All slides are air dried
- IHC slides are cut at 4 µm
- Standard tissue placement is mid-high on slide
Newcomer supply BCL2 (B-cell lymphoma 2) control slides are for the positive immunohistochemical staining of BCL2
Staining Results:
BCL2 positive expression will stain brown cytoplasmic staining.
Nuclei will stain blue.
The enclosed positive control slides are intended to be used to verify histological techniques and reagent reactivity. These slides are to be used for the qualitative purpose of determining positive or negative results, and are not intended to be used for any quantitative purpose.
Frequently used to distinguish positive staining follicular lymphoma from negative reacting follicular hyperplasia.
Ordering information: All controls are cut in serial sections.
10 set = 10 unstained slides and 1 stained slide.
98 set = 98 unstained slides and 2 stained slides. (First & Last Slides are Stained)
Reactivity of IHC control slides are guaranteed for one year from the date of receipt and should be revalidated after one year to verify continued reactivity.
Please note the following: Allow 2-3 weeks for delivery for large sets. In human tissue unless otherwise indicated.
10 set = 10 unstained slides and 1 stained slide.
98 set = 98 unstained slides and 2 stained slides. (First & Last Slides are Stained)
Reactivity of IHC control slides are guaranteed for one year from the date of receipt and should be revalidated after one year to verify continued reactivity.
Please note the following: Allow 2-3 weeks for delivery for large sets. In human tissue unless otherwise indicated.