ContactUsLinkComponent
Thermal Printer DPU-S445, Eppendorf
Thermal Printer DPU-S445, Eppendorf
Catalog # 75875-042
Supplier:  Eppendorf
undefined
Thermal Printer DPU-S445, Eppendorf
Catalog # 75875-042
Supplier:  Eppendorf
Supplier Number:  6135010004
Specifications
  • Description:
    Thermal Printer DPU-S445
  • Cat. No.:
    75875-042
  • Supplier no.:
    6135010004
Specifications
About this item
The Thermal Printer DPU-S445 is a thermal line dot printer that has a high printing speed while producing a clear type face
  • Automatic print-out of results
  • Easy and direct connection via USB
  • Compact and lightweight
  • Easy paper operation
Thermal printer is lightweight (approximately 0.5 kg). Maximum printing speed is 90 mm/second. The number of dots per line is 832, with a resolution of 8 dots/mm.
This allows the printer to print out scanned images. As an alternative to using USB sticks, the automatic print-out function of the DPU-S445 printer can be used to quickly obtain results.