PCR Tube Strips and Cap Strips, Eppendorf
Original Eppendorf PCR tubes are manufactured according to the highest quality standards.
- Eight reaction tubes in strip-format
- Ideal for small sample volumes
- Easily adaptable for automation
- Sealable using flat or domed strip-lids
- PCR with medium sample throughput
- Flat Cap Strips are suitable for real-time PCR
Eppendorf cap strips provide easy and rapid sealing of Eppendorf twin.tec® PCR plates, PCR strips and other standard multi-well plates. Strips with eight microcaps for 0.1 and 0.2 ml wells. Caps are autoclavable.
These thin-walled polypropylene tubes ensure efficient and homogenous heat transfer. The purity grade PCR Clean is safeguarded by closed bags being inside an outer carton.
Certified free from human DNA, DNase, RNase and PCR inhibitors.
- Catalog No:
- 75874-706
- 75874-708
- 75874-710
- 75874-702
- 75874-704
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental Certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
Frequently Bought Together