RmesFect™ Stem Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
Certificates
About this item
RmesFect Stem Transfection Reagent is based on the TEE-technology specifically designed for mRNA transfection in Stem Cells with high efficiency and low toxicity.
- Highly efficient: achieve mRNA delivery in all cells
- Ready-to-use: no need for additional buffer
- Low nucleic acid amount: minimized toxicity
- Protects mRNA against degradation
- Compatible with any culture medium
- Serum compatible: Medium change not required
Delivery: Shipped at room temperatue (RmesFect Kit GFP control shipped in dry ice)
Packaging: Store at -20°C (RmesFect Kit GFP control store at -80°C)
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together