Cell Culture Flasks, Eppendorf
About this item
Eppendorf Cell Culture Flasks are optimized for long-term cultivation or large-scale expansion of cells, providing the best protection against contamination.
- ConvexAcces™ geometry of the neck
- High efficiency air filter technology
- Defined arrested position on plug-seal caps
- Anti-rolling cap with corrugation
- Direct surface identification on flask and cap
Unsurpassed protection of your cells from contaminants was one of the key requirements in developing the Eppendorf cell culture flasks.
The defined arrested position on plug-seal caps prevent undesired closing. Anti-rolling cap with corrugation facilitates cultivation steps when the cap needs to be set aside. Can be centrifuged with the suitable adapters; refer to the manufacturer’s instruction. Pyrogen-, DNase-, RNase-, human and bacterial DNA-free. Non-cytotoxic.
A new, high efficiency air filter technology combines excellent protection with reliable gas exchange. In-line pressure testing for each flask guarantees leakage-free flasks for maximum workflow safety. The unique ConvexAccess™ shape of the neck facilitates the access to the growth area and makes cell seeding and media exchange much more convenient, safe, and reliable.
0 Options Available Below
Product Details & Documents
Eppendorf Cell Culture Flasks are optimized for long-term cultivation or large-scale expansion of cells, providing the best protection against contamination.
- ConvexAcces™ geometry of the neck
- High efficiency air filter technology
- Defined arrested position on plug-seal caps
- Anti-rolling cap with corrugation
- Direct surface identification on flask and cap
Unsurpassed protection of your cells from contaminants was one of the key requirements in developing the Eppendorf cell culture flasks.
The defined arrested position on plug-seal caps prevent undesired closing. Anti-rolling cap with corrugation facilitates cultivation steps when the cap needs to be set aside. Can be centrifuged with the suitable adapters; refer to the manufacturer’s instruction. Pyrogen-, DNase-, RNase-, human and bacterial DNA-free. Non-cytotoxic.
A new, high efficiency air filter technology combines excellent protection with reliable gas exchange. In-line pressure testing for each flask guarantees leakage-free flasks for maximum workflow safety. The unique ConvexAccess™ shape of the neck facilitates the access to the growth area and makes cell seeding and media exchange much more convenient, safe, and reliable.
Documents
Eppendorf Cell Culture Consumables Flyer
Product Literature
Product Family Options