Order Entry
United States
ContactUsLinkComponent
QMS™ Therapeutic Drug Monitoring (TDM) Assays Kits, Thermo Scientific
QMS™ Therapeutic Drug Monitoring (TDM) Assays Kits, Thermo Scientific
Catalog # 10154-282
CAS Number:  
QMS™ Therapeutic Drug Monitoring (TDM) Assays Kits, Thermo Scientific
Catalog # 10154-282
Supplier Number:  0373910
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Amikacin Assay kit
  • Environmentally Preferable:
  • Cat. No.:
    10154-282
  • Storage temperature:
    +2…+8 °C
  • Kit size:
    2×19, 2×7 ml
  • Sample type:
    Plasma serum
  • Supplier no.:
    0373910

Specifications

About this item

Measure therapeutic drug levels in serum and plasma with the Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Assays

  • Excellent correlation to reference methods
  • Optimal sensitivity, precision and dynamic range
  • Low cross-reactivity with metabolites and commonly co-administered drugs
  • Low interference from endogenous substances

These liquid, ready-to-use assays are a convenient alternative to more expensive and time-consuming technologies. QMS (Quantitative Microsphere System) immunoassays have excellent precision and accuracy, optimized performance on a wide range of general chemistry analyzers and lot-to-lot consistency.

For In Vitro diagnostic use.