To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:LPS Extraction Kit
- Environmentally Preferable:
- Cat. No.:102408-112
- Storage temperature:+4 °C
- No. of reactions:100× 5 ml bacterial cultures
- Supplier no.:17141
Specifications
About this item
The LPS Extraction Kit provides a new approach to extracting LPS with higher yields, fewer steps and in far less time
- Takes only 60 minutes to extract
- Gives reproducibly high yields
- Based on a robust water/phenol/chloroform extraction technology
Lipopolysaccharides (LPS) is a major constituent of the outer membrane of gram-negative bacteria. Some of LPS is a disease-causing endotoxin; it is also important in classification of serum type. Commonly using method is the complicated hot phenol-water extraction that takes long period of time.
Based on a robust water/phenol/chloroform extraction technology, the LPS extraction kit is flexible enough to run small culture volumes (<5 mL) or large (up to 100 mL) with similar yields.