Difco™ TSA with Lecithin and Polysorbate 80, BD Life Sciences
Catalog # 90005-312
Supplier: BD
CAS Number:
Specifications
- Description:Tsa with Leci+Polsorb80
- Size:20 mL
- Cat. No.:90005-312
- Supplier no.:299928
Specifications
About this item
BBL™ Trypticase™ Soy Agar (TSA) with Lecithin and Polysorbate 80 sterile pack tubes are used for the detection and enumeration of microorganisms present on surfaces of sanitary importance.
- Detection and enumeration of microorganisms
- Surface monitoring
- Used in air sampling equipment
The contact-style tubes are also recommended for use in air sampling equipment such as the Surface Air System.
The Sterile Pack tubes are particularly useful for monitoring surfaces in clean rooms and other environmentally controlled areas.