ContactUsLinkComponent
Magnetic Stirrers with Hot Plate, Yamato
undefined
Magnetic Stirrers with Hot Plate, Yamato
  • Catalog No:
  • 76550-250
  • Model:
  • Motor: AC Shading
  • Plate Material:
  • Aluminium, Ceramic Coated
  • Plate Dimensions:
  • Ø126mm×6pcs
  • W×D×H:
  • 606×420×122 mm (outside)
  • Control Type:
  • TRIAC Input temperature control
Frequently Bought Together