Orders LinkContactUsLinkComponent
MycoAlert™ Mycoplasma Detection Kits, Lonza
MycoAlert™ Mycoplasma Detection Kits, Lonza
Supplier:   Lonza
MycoAlert™ Mycoplasma Detection Kits, Lonza
Supplier:  Lonza
Certificates
MycoAlert™ Mycoplasma Detection Kits, Lonza
MycoAlert™ Mycoplasma Detection Kits, Lonza
9 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
The MycoAlert™ PLUS Mycoplasma Detection Kit is the next generation Lonza MycoAlert™ Kit. It provides higher light output to allow for use with less sensitive plate luminometers or multifunctional readers and testing of unused media, media supplements or water.
  • Convenient & fast
  • Simply add two reagents to the culture supernatant, perform two luminescence readings and get your results in less than 20 minutes
  • Detects all common mollicute contaminations (Mycoplasma, Acholeplasma, Entomoplasma and Spiroplasma; except Ureaplasma)
  • Suited for tube luminometers, plate luminometers and multifunctional readers
The MycoAlert™ PLUS Assay is a selective biochemical test that exploits the activity of mycoplasmal enzymes which are found in all six of the main mycoplasma cell culture contaminants and the vast majority of 180 mycoplasma species, but are not present in eukaryotic cells
MycoAlert™ Assay Control Set (available separately)
The MycoAlert™ Assay Control Set includes a lyophilized positive control and assay buffer for reconstitution. The assay buffer also serves as a negative control. The positive and negative controls are designed for use with the MycoAlert™ PLUS Mycoplasma Detection Kit. It is recommended to always include the Control Set to ensure that the operator and all of the assay reagents are performing properly. The positive control is proprietary, but does NOT contain mycoplasma. The positive or negative control is simply substituted for a sample in the standard assay protocol. The positive control will give a ratio of >1 while the negative control will give a ratio of <1.
Viable mycoplasma in a test sample (cell supernatant or fresh media or supplements) are lysed and the enzymes react with the MycoAlert™ PLUS Substrate, catalyzing the conversion of ADP to ATP. The ATP is then transferred into a light signal via the luciferase enzyme in the MycoAlert™ PLUS Reagent. By measuring the level of ATP in a sample both before (read A) and after the addition of the MycoAlert™ PLUS Substrate (read B), a ratio can be obtained which is indicative of the presence or absence of mycoplasma.
Product Family Options
Product Information
  • No. of tests
    Availability
    Price
  • 75860-358
    MycoAlert™ Plus Mycoplasma Detection Kits
    MycoAlert™ Plus Mycoplasma Detection Kits
    10
    Log in for availability
    Log in for availability
    Each
    Each - $320.52
    $320.52
    Each
       $320.52
    Minimum order quantity is 1
    Supplier Number: LT07-701
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 75860-360
    MycoAlert™ Plus Mycoplasma Detection Kits
    MycoAlert™ Plus Mycoplasma Detection Kits
    30
    Log in for availability
    Log in for availability
    Each
    Each - $755.68
    $755.68
    Each
       $755.68
    Minimum order quantity is 1
    Supplier Number: LT07-703
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 75860-362
    MycoAlert™ Plus Mycoplasma Detection Kits
    MycoAlert™ Plus Mycoplasma Detection Kits
    50
    Log in for availability
    Log in for availability
    Each
    Each - $1,281.30
    $1,281.30
    Each
       $1,281.30
    Minimum order quantity is 1
    Supplier Number: LT07-705
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 75866-212
    MycoAlert™ Plus Mycoplasma Detection Kits
    MycoAlert™ Plus Mycoplasma Detection Kits
    100
    Log in for availability
    Log in for availability
    Each
    Each - $1,617.12
    $1,617.12
    Each
       $1,617.12
    Minimum order quantity is 1
    Supplier Number: LT07-710
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 75870-452
    MycoAlert™ Mycoplasma Detection Kits
    MycoAlert™ Mycoplasma Detection Kits
    10
    Log in for availability
    Log in for availability
    Each
    Each - $321.99
    $321.99
    Each
       $321.99
    Minimum order quantity is 1
    Supplier Number: LT07-118
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 75870-454
    MycoAlert™ Mycoplasma Detection Kits
    MycoAlert™ Mycoplasma Detection Kits
    25
    Log in for availability
    Log in for availability
    Each
    Each - $621.91
    $621.91
    Each
       $621.91
    Minimum order quantity is 1
    Supplier Number: LT07-218
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 75870-456
    MycoAlert™ Mycoplasma Detection Kits
    MycoAlert™ Mycoplasma Detection Kits
    100
    Log in for availability
    Log in for availability
    Each
    Each - $1,634.10
    $1,634.10
    Each
       $1,634.10
    Minimum order quantity is 1
    Supplier Number: LT07-318
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 75870-458
    MycoAlert™ Mycoplasma Detection Kits
    MycoAlert™ Mycoplasma Detection Kits
    50
    Log in for availability
    Log in for availability
    Each
    Each - $1,061.42
    $1,061.42
    Each
       $1,061.42
    Minimum order quantity is 1
    Supplier Number: LT07-418
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 75870-450
    MycoAlert™ Assay Control Set
    MycoAlert™ Assay Control Set
    10
    Log in for availability
    Log in for availability
    Each
    Each - $294.47
    $294.47
    Each
       $294.47
    Minimum order quantity is 1
    Supplier Number: LT07-518
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.