Magnesium, Ion Chromatography Standard Solution, Specpure®, Mg²⁺ 1000 µg/ml, Thermo Scientific
Catalog # AA47258-AE
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Concentration:1000 µg/ml
- Matrix:H₂O
- Pack type:Glass Bottle
- Size:100 ml
- Cat. No.:AA47258-AE
- Element:Magnesium
- Supplier no.:47258-AE
Specifications
About this item
Magnesium nitrate hexahydrate in water