Calcium, Ion Chromatography Standard Solution, Specpure®, Ca²⁺ 1000 ug/ml, Thermo Scientific
Catalog # AA47234-AE
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Concentration:1000 µg/ml
- Matrix:H₂O
- Pack type:Glass Bottle
- Size:100 ml
- Cat. No.:AA47234-AE
- Element:Calcium
- Supplier no.:47234-AE
Specifications
About this item
Solution of calcium nitrate in water