Order Entry
United States
ContactUsLinkComponent
Nitroglycerin 1% in propylene glycol USP Reference Standard
Flammable
Toxic
Health
Irritant
Nitroglycerin 1% in propylene glycol USP Reference Standard
Catalog # 101095-632
Supplier:  USP
CAS Number:  
Flammable
Toxic
Health
Irritant
Nitroglycerin 1% in propylene glycol USP Reference Standard
Catalog # 101095-632
Supplier:  USP
Supplier Number:  1466506
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Ampoule
  • Size:
    5×200 mg
  • Cat. No.:
    101095-632
  • Supplier no.:
    1466506

Specifications

About this item

 :
 :227.085 g/mol
UN:1204
ADR:3,II
 :GHS02
 :GHS06
 :GHS08
 :GHS07
CAS Number:55-63-0
Formula:C₃H5N₃O₉