ContactUsLinkComponent
EnzyChrom™ Triglyceride Assay Kit, BioAssay Systems
Catalog # 75878-176
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
EnzyChrom™ Triglyceride Assay Kit, BioAssay Systems
Catalog # 75878-176
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  ETGA-200
CAS Number:  
Specifications
  • Description:
    Triglyceride Assay Kit
  • Size:
    200 assays
  • Method:
    OD570 nm
  • Sample:
    Serum, plasma, etc.
  • Processing Time:
    30 minutes
  • Cat. No.:
    75878-176
  • Supplier no.:
    ETGA-200
Specifications
About this item
For quantitative determination of triglyceride and evaluation of drug effects on triglyceride metabolism.
  • Linear detection range 0.01 - 1.0 mmol/L (0.88 - 88.5 mg/dL) triglyceride
  • Convenient room temperature high-throughput assay
Triglyceride, also known as Triacyltriglyceride or Triacyl-glyceride, is the main constituent in vegetable oil and animal fats
BioAssay Systems’ triglyceride assay uses a single Working Reagent that combines triglyceride hydrolysis and glycerol determination in one step, in which a dye reagent is oxidized to form a colored product. The color intensity at 570nm is directly proportional to triglyceride concentration in the sample.
Triglycerides play an important role as energy sources and transporters of dietary fat. In the human body, high levels of triglycerides in the bloodstream have been linked to atherosclerosis, heart disease and pancreatitis. Simple, direct and automation-ready. Procedures for measuring triglyceride concentrations find wide applications in research and drug discovery.