Orders LinkContactUsLinkComponent
Labtainer™ BioProcess Containers, 3 Ports, Thermo Scientific
Labtainer™ BioProcess Containers, 3 Ports, Thermo Scientific
Certificates
Labtainer™ BioProcess Containers, 3 Ports, Thermo Scientific
20 Options Available Below
Product Details & Documents
Small volume liquid handling systems for cell culture and bioprocessing.
  • Space saving and ergonomic
  • Dispensing, packaging and storing cell culture media, buffers and process liquids
  • Delivery of cell culture media or process liquids to small scale bioproduction systems
  • Bioreactor and fermentation feed, sampling and harvest
  • Storage and transport of bulk intermediate products, process intermediates, vaccine conjugates and other biological products
They range in size from 50 ml to 50 L, with a variety of standard configurations to meet most application needs.
 : Constructed in an ISO 7 cleanroom under cGMP conditions.
Product Family Options
Product Information
  • Capacity
    Packaging
    Port 1
    Port 2
    Port 3
    W×L
    Availability
    Price
  • 2 L
    Single pack
    C flex tubing, connection: 0.64×0.97 cm (0.25×0.38") length: 30 cm (12"), Luer lock insert, polypropylene
    C flex tubing, connection: 0.64×0.97 cm (0.25×0.38") length: 30 cm (12"), Luer lock insert, polypropylene
    C flex tubing, connection: 0.32×0.64 cm (0.13×0.25") length: 30 cm (12"), Luer lock body, polypropylene
    310×295 mm (12.2×11.6")
    Log in for availability
    Log in for availability
    Each (2l)
    Each (2l) - $126.17
    $126.17
    New Customer Promotional Price
    Each (2l)
       $126.17
    Supplier Number: SH30713.05
  • 5 L
    Single pack
    C flex tubing, connection: 0.64×0.97 cm (0.25×0.38") length: 30 cm (12"), Luer lock insert, polypropylene
    C flex tubing, connection: 0.64×0.97 cm (0.25×0.38") length: 30 cm (12"), Luer lock insert, polypropylene
    C flex tubing, connection: 0.32×0.64 cm (0.13×0.25") length: 30 cm (12"), Luer lock body, polypropylene
    333×376 mm (13.1×14.8")
    Log in for availability
    Log in for availability
    Each (5l)
    Each (5l) - $125.47
    $125.47
    New Customer Promotional Price
    Each (5l)
       $125.47
    Supplier Number: SH30713.01
  • 10 L
    Single pack
    C flex tubing, connection: 0.64×0.97 cm (0.25×0.38") length: 30 cm (12"), Luer lock insert, polypropylene
    C flex tubing, connection: 0.64×0.97 cm (0.25×0.38") length: 30 cm (12"), Luer lock insert, polypropylene
    C flex tubing, connection: 0.32×0.64 cm (0.13×0.25") length: 30 cm (12"), Luer lock body, polypropylene
    300×622 mm (11.8×24.5")
    Log in for availability
    Log in for availability
    Each (10l)
    Each (10l) - $122.43
    $122.43
    New Customer Promotional Price
    Each (10l)
       $122.43
    Supplier Number: SH30713.02
  • 20 L
    Single pack
    C flex tubing, connection: 0.64×0.97 cm (0.25×0.38") length: 30 cm (12"), Luer lock insert, polypropylene
    C flex tubing, connection: 0.64×0.97 cm (0.25×0.38") length: 30 cm (12"), Luer lock insert, polypropylene
    C flex tubing, connection: 0.32×0.64 cm (0.13×0.25") length: 30 cm (12"), Luer lock body, polypropylene
    432×655 mm (17×25.8")
    Log in for availability
    Log in for availability
    Each (20l)
    Each (20l) - $132.25
    $132.25
    New Customer Promotional Price
    Each (20l)
       $132.25
    Supplier Number: SH30713.03
  • 50 L
    Single pack
    C flex tubing, connection: 0.64×0.97 cm (0.25×0.38") length: 30 cm (12"), Luer lock insert, polypropylene
    C flex tubing, connection: 0.64×0.97 cm (0.25×0.38") length: 30 cm (12"), Luer lock insert, polypropylene
    C flex tubing, connection: 0.32×0.64 cm (0.13×0.25") length: 30 cm (12"), Luer lock body, polypropylene
    584×826 mm (23×32.5")
    Log in for availability
    Log in for availability
    Each (50l)
    Each (50l) - $190.56
    $190.56
    New Customer Promotional Price
    Each (50l)
       $190.56
    Supplier Number: SH30713.04
  • 2 L
    Single pack
    C flex tubing, connection: 0.97×1.59 cm (0.38×0.63") length: 61 cm (24"), MPC insert, polycarbonate
    C flex tubing, connection: 0.97×1.59 cm (0.38×0.63") length: 61 cm (24"), MPC insert, polycarbonate
    C flex tubing, connection: 0.32×0.64 cm (0.13×0.25") length: 61 cm (24"), Luer lock body connection, polypropylene
    310×295 mm (12.2×11.6")
    Log in for availability
    Log in for availability
    Each (2l)
    Each (2l) - $89.61
    $89.61
    New Customer Promotional Price
    Each (2l)
       $89.61
    Supplier Number: SH30712.05
  • 5 L
    Single pack
    C flex tubing, connection: 0.97×1.59 cm (0.38×0.63") length: 61 cm (24"), MPC insert, polycarbonate
    C flex tubing, connection: 0.97×1.59 cm (0.38×0.63") length: 61 cm (24"), MPC insert, polycarbonate
    C flex tubing, connection: 0.32×0.64 cm (0.13×0.25") length: 61 cm (24"), Luer lock body connection, polypropylene
    333×376 mm (13.1×14.8")
    Log in for availability
    Log in for availability
    Each (5l)
    Each (5l) - $117.27
    $117.27
    New Customer Promotional Price
    Each (5l)
       $117.27
    Supplier Number: SH30712.01
  • 10 L
    Single pack
    C flex tubing, connection: 0.97×1.59 cm (0.38×0.63") length: 61 cm (24"), MPC insert, polycarbonate
    C flex tubing, connection: 0.97×1.59 cm (0.38×0.63") length: 61 cm (24"), MPC insert, polycarbonate
    C flex tubing, connection: 0.32×0.64 cm (0.13×0.25") length: 61 cm (24"), Luer lock body connection, polypropylene
    300×622 mm (11.8×24.5")
    Log in for availability
    Log in for availability
    Each (10l)
    Each (10l) - $119.97
    $119.97
    New Customer Promotional Price
    Each (10l)
       $119.97
    Supplier Number: SH30712.02
  • 20 L
    Single pack
    C flex tubing, connection: 0.97×1.59 cm (0.38×0.63") length: 61 cm (24"), MPC insert, polycarbonate
    C flex tubing, connection: 0.97×1.59 cm (0.38×0.63") length: 61 cm (24"), MPC insert, polycarbonate
    C flex tubing, connection: 0.32×0.64 cm (0.13×0.25") length: 61 cm (24"), Luer lock body connection, polypropylene
    432×655 mm (17×25.8")
    Log in for availability
    Log in for availability
    Each (20l)
    Each (20l) - $140.89
    $140.89
    New Customer Promotional Price
    Each (20l)
       $140.89
    Supplier Number: SH30712.03
  • 50 L
    Single pack
    C flex tubing, connection: 0.97×1.59 cm (0.38×0.63") length: 61 cm (24"), MPC insert, polycarbonate
    C flex tubing, connection: 0.97×1.59 cm (0.38×0.63") length: 61 cm (24"), MPC insert, polycarbonate
    C flex tubing, connection: 0.32×0.64 cm (0.13×0.25") length: 61 cm (24"), Luer lock body connection, polypropylene
    584×826 mm (23×32.5")
    Log in for availability
    Log in for availability
    Each (50l)
    Each (50l) - $223.51
    $223.51
    New Customer Promotional Price
    Each (50l)
       $223.51
    Supplier Number: SH30712.04
  • 2 L
    Single pack
    C flex tubing, connection: 0.97×1.27 cm (0.38×0.5") length: 30 cm (12"), MPC insert, polycarbonate
    C flex tubing, connection: 0.97×1.27 cm (0.38×0.5") length: 30 cm (12"), MPC insert, polycarbonate
    No tubing, plugged
    310×295 mm (12.2×11.6")
    Log in for availability
    Log in for availability
    Each (2l)
    Each (2l) - $130.77
    $130.77
    New Customer Promotional Price
    Each (2l)
       $130.77
    Supplier Number: SH30714.05
  • 5 L
    Single pack
    C flex tubing, connection: 0.97×1.27 cm (0.38×0.5") length: 30 cm (12"), MPC insert, polycarbonate
    C flex tubing, connection: 0.97×1.27 cm (0.38×0.5") length: 30 cm (12"), MPC insert, polycarbonate
    No tubing, plugged
    333×376 mm (13.1×14.8")
    Log in for availability
    Log in for availability
    Each (5l)
    Each (5l) - $97.09
    $97.09
    New Customer Promotional Price
    Each (5l)
       $97.09
    Supplier Number: SH30714.01
  • 10 L
    Single pack
    C flex tubing, connection: 0.97×1.27 cm (0.38×0.5") length: 30 cm (12"), MPC insert, polycarbonate
    C flex tubing, connection: 0.97×1.27 cm (0.38×0.5") length: 30 cm (12"), MPC insert, polycarbonate
    No tubing, plugged
    300×622 mm (11.8×24.5")
    Log in for availability
    Log in for availability
    Each (10l)
    Each (10l) - $103.11
    $103.11
    New Customer Promotional Price
    Each (10l)
       $103.11
    Supplier Number: SH30714.02
  • 20 L
    Single pack
    C flex tubing, connection: 0.97×1.27 cm (0.38×0.5") length: 30 cm (12"), MPC insert, polycarbonate
    C flex tubing, connection: 0.97×1.27 cm (0.38×0.5") length: 30 cm (12"), MPC insert, polycarbonate
    No tubing, plugged
    432×655 mm (17×25.8")
    Log in for availability
    Log in for availability
    Each (20l)
    Each (20l) - $109.80
    $109.80
    New Customer Promotional Price
    Each (20l)
       $109.80
    Supplier Number: SH30714.03
  • 50 L
    Single pack
    C flex tubing, connection: 0.97×1.27 cm (0.38×0.5") length: 30 cm (12"), MPC insert, polycarbonate
    C flex tubing, connection: 0.97×1.27 cm (0.38×0.5") length: 30 cm (12"), MPC insert, polycarbonate
    No tubing, plugged
    584×826 mm (23×32.5")
    Log in for availability
    Log in for availability
    Each (50l)
    Each (50l) - $185.68
    $185.68
    New Customer Promotional Price
    Each (50l)
       $185.68
    Supplier Number: SH30714.04
  • 2 L
    Single pack, Edge ports
    No tubing, MPC insert, polycarbonate
    No tubing, MPC insert, polycarbonate
    No tubing, injection port
    310×295 mm (12.2×11.6")
    Log in for availability
    Log in for availability
    Each (2l)
    Each (2l) - $76.75
    $76.75
    New Customer Promotional Price
    Each (2l)
       $76.75
    Supplier Number: SH30709.05
  • 5 L
    Single pack, Edge ports
    No tubing, MPC insert, polycarbonate
    No tubing, MPC insert, polycarbonate
    No tubing, injection port
    333×376 mm (13.1×14.8")
    Log in for availability
    Log in for availability
    Each (5l)
    Each (5l) - $87.14
    $87.14
    New Customer Promotional Price
    Each (5l)
       $87.14
    Supplier Number: SH30709.01
  • 10 L
    Single pack, Edge ports
    No tubing, MPC insert, polycarbonate
    No tubing, MPC insert, polycarbonate
    No tubing, injection port
    300×622 mm (11.8×24.5")
    Log in for availability
    Log in for availability
    Each (10l)
    Each (10l) - $89.33
    $89.33
    New Customer Promotional Price
    Each (10l)
       $89.33
    Supplier Number: SH30709.02
  • 20 L
    Single pack, Edge ports
    No tubing, MPC insert, polycarbonate
    No tubing, MPC insert, polycarbonate
    No tubing, injection port
    432×655 mm (17×25.8")
    Log in for availability
    Log in for availability
    Each
    Each - $96.11
    $96.11
    New Customer Promotional Price
    Each
       $96.11
    Supplier Number: SH30709.03
  • 50 L
    Single pack, Edge ports
    No tubing, MPC insert, polycarbonate
    No tubing, MPC insert, polycarbonate
    No tubing, injection port
    584×826 mm (23×32.5")
    Log in for availability
    Log in for availability
    Each (50l)
    Each (50l) - $190.73
    $190.73
    New Customer Promotional Price
    Each (50l)
       $190.73
    Supplier Number: SH30709.04