EnzyChrom™ Glutathione Peroxidase Assay Kit, BioAssay Systems
Catalog # 75878-136
Supplier: BIOASSAY SYSTEMS
CAS Number:
Specifications
- Description:Glutathione Peroxidase Assay Kit
- Size:100 assays
- Method:OD340 nm
- Sample:Biological
- Processing Time:20 minutes
- Cat. No.:75878-136
- Supplier no.:EGPX-100
Specifications
About this item
For quantitative determination of glutathione peroxidase activity.
- Linear detection range 40 to 800 U/L GPX activity
Glutathione peroxidase (GPX, EC 1.11.1.9) represents an enzyme family with peroxidase activity whose main biological role is to protect the organism from oxidative damage
BioAssay Systems’ improved assay directly measures NADPH consumption in the enzyme coupled reactions. The measured decrease in optical density at 340 nm is directly proportional to the enzyme activity in the sample.
It helps prevent lipid peroxidation of cellular membranes by removing free peroxide in the cell. GPX catalyzes the following reaction with glutathione reductase (GR), Simple, direct and high-throughput assays for GPX activity find wide applications.