ContactUsLinkComponent
EnzyChrom™ Glucose-6-Phosphate Assay Kit, BioAssay Systems
EnzyChrom™ Glucose-6-Phosphate Assay Kit, BioAssay Systems
Catalog # 75878-112
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
EnzyChrom™ Glucose-6-Phosphate Assay Kit, BioAssay Systems
Catalog # 75878-112
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  EG6P-100
CAS Number:  
Specifications
  • Description:
    Glucose-6-Phosphate Assay Kit
  • Size:
    100 assays
  • Method:
    OD460 nm
  • Sample:
    Plasma, serum, tissue and culture media etc.
  • Processing Time:
    20 minutes
  • Cat. No.:
    75878-112
  • Supplier no.:
    EG6P-100
Specifications
About this item
For quantitative determination of glucose-6-phosphate in biological samples.
  • Linear detection range 10 to 1000 μM G6P
  • Homogeneous "mix-incubate-measure" type high-throughput assay
Glucose-6-Phosphate (g6p) is glucose sugar phosphorylated on carbon 6
BioAssay Systems’ Glucose-6-Phosphate Assay Kit provides a simple, and automation-ready procedure for measuring G6P concentration. G6P is oxidized by glucose-6-phosphate dehydrogenase and the formed NADPH is coupled to the formazan (WST-8) chromogen. The intensity of the product color, measured at 460 nm, is proportional to the G6P concentration in the sample.
Most of the glucose entering cells is phosphorylated to G6P. G6P has three primary fates within the cell. It lies at the start of two major metabolic pathways: glycolysis and the pentose phosphate pathway. In addition to these metabolic pathways, glucose 6-phosphate may also be converted to glycogen or starch for storage.