Specifications
- Description:Aspartate Transaminase Assay Kit
- Size:100 assays
- Method:OD340 nm
- Sample:Serum, plasma, etc.
- Processing Time:10 minutes
- Cat. No.:75878-048
- Supplier no.:EASTR-100
Specifications
About this item
For quantitative determination of aspartate transaminase (AST) activity and evaluation of drug effects on AST activity.
- Linear detection range 2-100 U/L
- Assay can be carried out in a microplate or a cuvette and takes only 10 minutes
Aspartate Transaminase (AST), also known as serum glutamic oxaloacetic transaminase (GOT) or aspartate aminotransferase (ASAT/AAT), facilitates the conversion of aspartate and a-ketoglutarate to oxaloacetate and glutamate
BioAssay Systems’ AST activity assay is based on the quantification of oxaloacetate produced by AST. In this assay, oxaloacetate and NADH are converted to malate and NAD by the enzyme malate dehydrogenase. The decrease in NADH absorbance at 340 nm is proportionate to AST activity.
There are two isoenzymes in humans: GOT1 is a cytosolic isoenzyme derived from red blood cells and heart; GOT2 is the mitochondrial isoenzyme found mainly in the liver. AST is elevated in liver and muscle diseases. It is part of diagnostic tests for liver function, myocardial infarction, acute pancreatitis, acute hemolytic anemia, severe burns, acute renal disease and trauma. Simple, direct and automation-ready. Procedures for measuring AST activity find wide applications in research and drug discovery.