ContactUsLinkComponent
MicroFlow™ PFA Self-Aspirating Nebulizers, PerkinElmer
MicroFlow™ PFA Self-Aspirating Nebulizers, PerkinElmer
Catalog # 10136-716
Supplier:  PerkinElmer
undefined
MicroFlow™ PFA Self-Aspirating Nebulizers, PerkinElmer
Catalog # 10136-716
Supplier:  PerkinElmer
Supplier Number:  N8145102
Specifications
  • Description:
    PFA Nebulizers with Integrated Fluoropolymer Capillaries, 50 μL/min
  • Cat. No.:
    10136-716
  • Supplier no.:
    N8145102
Specifications
About this item
An integrated capillary PFA nebulizer for self aspiration.
  • Resistant to clogging
  • Produces a fine aerosol for high transport efficiency and high sensitivity
  • Low, spike free background for important elements such as Fe and Ca
  • Direct analysis of volatile and non-volatile organic solvents
PFA MicroFlow nebulizers are constructed entirely from chemically resistant fluoropolymers, ideal for strong acids, alkalis, organics and high salt solutions.