Specifications
- Description:TBARS (Lipid Peroxide) Assay Kit
- Size:100 assays
- Method:OD535 nm, or FL560/585 nm
- Sample:Serum, plasma, urine, saliva etc
- Processing Time:80 minutes
- Cat. No.:75878-012
- Supplier no.:DTBA-100
Specifications
About this item
For quantitative determination of lipid peroxides (thiobarbituric acid reactive substances, TBARS).
- Linear detection range colorimetric assay 1 - 30 μM and fluorometric assay 0.1 - 1.5 μM MDA
Oxidative attack of essential cell components by reactive oxygen species has been associated with several human diseases, such as atherosclerosis, cardiovascular diseases, diabetes, liver disorders, and inflammatory rheumatic diseases
BioAssay Systems’ TBARS assay is based on the reaction of TBARS with thiobarbituric acid (TBA) to form a pink colored product. The color intensity at 535nm or fluorescence intensity at (λex/em = 560nm/585nm) is directly proportional to TBARS concentration in the sample.
THIOBARBITURIC ACID REACTIVE SUBSTANCES (TBARS) are low-molecular-weight end products (mainly malondialdehyde, MDA) that are formed during the decomposition of lipid peroxidation products. Increased levels of TBARS have been demonstrated in these diseases. Simple, direct and accurate assays for TBARS find wide applications in research and drug discovery.