ContactUsLinkComponent
Glucose Assay Kit , BioAssay Systems
Glucose Assay Kit , BioAssay Systems
Catalog # 75878-100
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
Glucose Assay Kit , BioAssay Systems
Catalog # 75878-100
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  DIGL-100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Glucose Assay Kit
  • Size:
    100 assays
  • Method:
    OD630 nm (Chemical)
  • Sample:
    Biological (e.g. serum, plasma), food and beverage.
  • Processing Time:
    10 minutes
  • Brand Name:
    QuantiChrom™
  • Cat. No.:
    75878-100
  • Supplier no.:
    DIGL-100
Specifications
About this item
For quantitative determination of glucose and evaluation of drug effects on glucose metabolism
  • Sensitive and accurate. Use as little as 5 μL samples. Linear detection range 0.7 mg/dL (39 μM) to 300 mg/dL (16.6 mM) glucose in 96-well plate.
  • Simple and convenient. The procedure involves addition of a single working reagent and incubation for 8 min in a boiling water bath.
  • Improved reagent stability. The optimized formulation has greatly enhanced the reagent and signal stability.
  • Low interference in biological samples. No pretreatments are needed. Assays can be directly performed on serum and plasma samples.
  • Method: OD630nm (Chemical)
  • Samples: Biological, food and beverage
Glucose (C6H12O6) is a ubiquitous fuel molecule in biology. It is oxidized through a series of enzyme-catalyzed reactions to form carbon dioxide and water, yielding the universal energy molecule ATP. Due to its importance in metabolism, glucose level is a key diagnostic parameter for many metabolic disorders. Increased glucose levels have been associated with diabetes mellitus, hyperactivity of thyroid, pituitary and adrenal glands. Decreased levels are found in insulin secreting tumors, myxedema, hypopituitarism and hypoadrenalism. Simple, direct and automation-ready procedures for measuring glucose concentrations find wide applications in research and drug discovery. BioAssay Systems’ glucose assay kit is designed to measure glucose directly in serum or plasma without any pretreatment. The improved o-toluidine method utilizes a specific color reaction with glucose. The absorbance at 630nm is directly proportional to glucose concentration in the sample.