ContactUsLinkComponent
Isocitrate Dehydrogenase Assay Kit, BioAssay Systems
Isocitrate Dehydrogenase Assay Kit, BioAssay Systems
Catalog # 75878-094
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
Isocitrate Dehydrogenase Assay Kit, BioAssay Systems
Catalog # 75878-094
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  DIDH-100
CAS Number:  
Specifications
  • Description:
    Isocitrate Dehydrogenase Assay Kit
  • Size:
    100 assays
  • Method:
    OD565 nm
  • Sample:
    Plasma, serum,tissue and culture media etc.
  • Processing Time:
    30 minutes
  • Brand Name:
    QuantiChrom™
  • Cat. No.:
    75878-094
  • Supplier no.:
    DIDH-100
Specifications
About this item
For quantitative colorimetric kinetic determination of isocitrate dehydrogenase activity and evaluation of drug effects on its metabolism
  • Fast and sensitive. Linear detection range (50µL sample): 0.5 µM to 60 µM acetaldehyde in 96-well plate assay.
  • Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay.
  • Can be readily automated on HTS liquid handling systems for processing thousands of samples per day.
  • Method: OD565nm
  • Samples: Plasma, serum,tissue and culture media etc
ISOCITRATE DEHYDROGENASE (IDH) is an enzyme which catalyzes the interconversion of isocitrate and a-ketoglutarate. There are three IDH isoforms: IDH3 uses the cofactor NAD+ and catalyzes the third step in the citric acid cycle, while IDH1 and IDH2 use the cofactor NADP+ and catalyze the same reaction outside the citric acid cycle. This kit measures the activity of the NADP+ isoforms. Mutations in IDH1 and IDH2 have been linked with various brain tumors and acute myeloid leukemia. BioAssay Systems’ non-radioactive, colorimetric IDH assay is based on the reduction of the tetrazolium salt MTT in a NADPH-coupled enzymatic reaction to a reduced form of MTT which exhibits an absorption maximum at 565 nm. The increase in absorbance at 565 nm is directly proportional to the enzyme activity.