ContactUsLinkComponent
QuantiFluo™ Caspase Assay Kit, BioAssay Systems
QuantiFluo™ Caspase Assay Kit, BioAssay Systems
Catalog # 75877-968
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
QuantiFluo™ Caspase Assay Kit, BioAssay Systems
Catalog # 75877-968
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  DCS3-100
CAS Number:  
Specifications
  • Description:
    Caspase Assay Kit
  • Size:
    100 assays
  • Method:
    FL400/490 nm
  • Sample:
    Cell and tissue lysate
  • Processing Time:
    60 minutes
  • Cat. No.:
    75877-968
  • Supplier no.:
    DCS3-100
Specifications
About this item
For quantitative determination of the apoptosis target caspase-3 activity and HTS screen for apoptosis inducers and inhibitors.
  • Safe
  • Non-radioactive assay
  • This homogeneous "mix-incubate-measure" type assay can be readily automated for processing thousands of samples per day
Caspases are members of the aspartate-specific cysteinyl protease family that play a central role in apoptosis
BioAssay Systems’ Caspase-3 Assay Kit provides a convenient means to measure caspase-3 activity in biological samples. In the assay, a specific substrate (N-Ac-DEVD-AFC) is cleaved by active caspase-3, forming a highly fluorescent product. The fluorescence intensity (λex/em = 400/490nm) is proportional to the caspase-3 activity.
Apoptosis is involved in a variety of physiological and pathological events, ranging from normal fetal development to diseases such as cancer, organ failure, and neurodegenerative diseases. Caspase-3 is key biomarker in the assessment of apoptosis and in understanding mechanism of apoptosis induction.