Anti-SYP Rabbit Monoclonal Antibody [clone: RM258]
Catalog # 103320-668
Supplier: RevMAb Biosciences
Specifications
- Antibody Type:Primary
- Antigen Name:Synaptophysin
- Antigen Symbol:SYP
- Clonality:Monoclonal
- Clone:RM258
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µL
- Epitope:C-terminal
- Format:Purified
- Form:Liquid
- Gene ID:6855
- Storage Buffer:50% Glycerol/PBS with 1% BSA and 0.09% sodium azide
- Storage Temperature:–20.0 °C
- Shipping Temperature:4°C
- Immunogen:A peptide corresponding to the C-terminus of Synaptophysin
- Purification:Protein A affinity purified from an animal origin-free culture supernatant
- Cat. No.:103320-668
- Supplier no.:31-1139-00
Specifications
About this item
This antibody reacts to human Synaptophysin.
Western Blot (WB): 1:1000-1:2000 dilution;
Immunohistochemistry (IHC): 1:200- 1:500 dilution
Type: Primary
Antigen: SYP
Clonality: Monoclonal
Clone: RM258
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human