Specifications
- Description:DREL Complete Water Quality Lab
- Model:DR 1900
- Battery Life:15 days (typical)
- Wavelength Range:340 – 800 nm
- Wavelength Accuracy:±2 nm
- Operating Humidity:max. 80 %
- Photometric Measuring Range:0 - 3 Abs
- Interfaces:USB
- Operating Temperature:0…50 °C
- Optics:Reference beam, spectral, Silicon photodiode detector
- Power supply:4x AA battery
- Weight:1.5 kg
- Cat. No.:75875-618
- Range:DR 1900: Wavelength range 340 nm - 800 nm
- Supplier no.:2922401
Specifications
About this item
Hach DREL Portable Water Quality Laboratories are for customers that need flexibility, accuracy and convenience of a portable spectrophotometer, with all necessary reagents and apparatus, in a rugged, well-organized carrying case
- Contains durable instruments, premeasured reagents impervious to environmental contamination, and apparatus and labware
- Instruments have the option of running on batteries, so tests can be run without concern for sample preservation and handling
- Rugged kit is designed to withstand harsh field conditions
- All-new DR1900 portable spectrophotometer is incredibly light (3.3 lbs) with battery, and can read Hach TNT+ vials
Contains a DR 1900 portable spectrophotometer and Digital Titrator, along with reagents and apparatus necessary to run approximately 100 tests on 24 different parameters. Two test kit cases with molded inserts are included. One case holds instrument and accessories, the other holds reagents and testing apparatus. The instrument case has additional storage space for adding optional pH meter, portable turbimeter, and 2 probes.
Case size (2 cases): 21x11.5x11 (WxDxH)
These kits all include a DR 1900, reagents and apparatus necessary for running each included test 100 times