Limabean Agar, Himedia
Catalog # 61000-180
Supplier: HiMedia
CAS Number:
Specifications
- Size:500 g
- Cat. No.:61000-180
- Supplier no.:M736-500G
Specifications
About this item
For the cultivation of phytopathological and other fungi.
For the cultivation of phytopathological and other fungi.