ContactUsLinkComponent
Balston Type 95A Acetylene Filter, PerkinElmer
Balston Type 95A Acetylene Filter, PerkinElmer
  97013-124
undefined
Balston Type 95A Acetylene Filter, PerkinElmer
  97013-124
 :  N9301399
  • Description:
    Acetylene Filter
  • Cat. No.:
    97013-124
  • Supplier no.:
    N9301399
Filters remove particulate and acetone droplets from acetylene to 99.99% at 0.1 µm, protecting microcomputer gas controls and AA burner systems from contamination and corrosion.
Recommended for use with AA spectrometers, where the air/acetylene filter is not installed. Includes mounting bracket and appropriate fittings for installation on an acetylene line.